Tisk…

2,4-Dimethylthiazole-5-sulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR23081-1G OR23081-250MG
APOSOR23081-1GEA 4100 CZK
APOSOR23081-1G APOSOR23081-250MG
2,4-Dimethylthiazole-5-sulphonyl chloride
2,4-Dimethylthiazole-5-sulphonyl chloride
Vzorec: C₅H₆ClNO₂S₂
Bod varu: 82…84 °C/ 0,3mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00052746
CAS číslo: 80466-80-4
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR