Tisk…

Isonicotinoyl chloride hydrochloride 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Pyridine-4-carbonyl Chloride Hydrochloride, 4-Pyridinoyl chloride hydrochloride

OR23065-5G OR23065-25G
APOSOR23065-5GEA 622 CZK
APOSOR23065-5G APOSOR23065-25G
Isonicotinoyl chloride hydrochloride 95%
Isonicotinoyl chloride hydrochloride
Vzorec: C₆H₄ClNO
MW: 178,01 g/mol
Bod tání: 153…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00012830
CAS číslo: 39178-35-3
EINECS: 254-331-2
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR