Picolinic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Pyridine-2-carboxylic acid, 2-Picolinic acid, 2-Pyridinecarboxylic acid
OR23054-5G
OR23054-500G
OR23054-1KG
APOSOR23054-5GEA
622
CZK
APOSOR23054-5G
APOSOR23054-500G
APOSOR23054-1KG
Picolinic acid
Picolinic acid
Vzorec:
C₆H₅NO₂ MW: 123,11 g/mol Bod tání: 139…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006293 CAS číslo: 98-98-6 EINECS: 202-719-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...