4-Bromo-2-chlorobenzenesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-Bromo-2-chlorobenzene-1-sulphonyl chloride
OR2305-25G
OR2305-5G
APOSOR2305-25GEA
1580
CZK
APOSOR2305-25G
APOSOR2305-5G
4-Bromo-2-chlorobenzenesulphonyl chloride
4-Bromo-2-chlorobenzenesulphonyl chloride
Vzorec:
C₆H₃BrCl₂O₂S MW: 289,96 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03094630 CAS číslo: 351003-52-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...