2,3-Dihydrobenzo[b][1,4]dioxine-5-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,3-dihydro-1,4-benzodioxine-5-carboxylic acid, 2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylic acid, 1,4-Benzodioxan-5-carboxylic acid
OR23013-1G
OR23013-10G
OR23013-25G
APOSOR23013-1GEA
622
CZK
APOSOR23013-1G
APOSOR23013-10G
APOSOR23013-25G
2,3-Dihydrobenzo[b][1,4]dioxine-5-carboxylic acid
2,3-Dihydrobenzo[b][1,4]dioxine-5-carboxylic acid
Vzorec:
C₉H₈O₄ MW: 180,16 g/mol Bod tání: 90…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00239415 CAS číslo: 4442-53-9 EINECS: 224-670-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...