3,4-(Ethylenedioxy)benzylamine 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
6-(Aminomethyl)-2,3-dihydro-1,4-benzodioxine, (2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)methanamine
OR23012-1G
OR23012-5G
APOSOR23012-1GEA
2860
CZK
APOSOR23012-1G
APOSOR23012-5G
3,4-(Ethylenedioxy)benzylamine 95%
3,4-(Ethylenedioxy)benzylamine
Vzorec:
C₈H₉NO₂ MW: 165,1891 g/mol Bod varu: 105…110 °C/0,5mmHg Storage Temperature: Ambient |
MDL Number:
MFCD02677684 CAS číslo: 17413-10-4 EINECS: 241-440-5 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...