Tisk…

tert-Butyl-N-(3-thienyl)carbamate

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl thiophen-3-ylcarbamate, 3-Aminothiophene, N-BOC protected

OR22965-1G OR22965-5G OR22965-10G
APOSOR22965-1GEA 953 CZK
APOSOR22965-1G APOSOR22965-5G APOSOR22965-10G
tert-Butyl-N-(3-thienyl)carbamate
tert-Butyl-N-(3-thienyl)carbamate
Vzorec: C₉H₁₃NO₂S
MW: 199,27 g/mol
Bod tání: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD01928808
CAS číslo: 19228-91-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR