2,3-Dihydrobenzo[b]furan-5-carbaldehyde 95+%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
5-Coumarancarboxaldehyde, 2,3-Dihydrobenzofuran-5-carbaldehyde, 2,3-Dihydrobenzo[b]furan-5-carboxaldehyde, 2,3-Dihydrobenzofuran-5-carboxaldehyde
OR2280-25G
OR2280-100G
APOSOR2280-25GEA
1870
CZK
APOSOR2280-25G
APOSOR2280-100G
2,3-Dihydrobenzo[b]furan-5-carbaldehyde 95+%
2,3-Dihydrobenzo[b]furan-5-carbaldehyde
Vzorec:
C₉H₈O₂ MW: 148,16 g/mol Bod varu: 88…89 °C/0,1mmHg Hustota: 1,177 g/cm³ (25 °C) Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068058 CAS číslo: 55745-70-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...