1-Phenylcyclobutanecarboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR2270-1G
OR2270-250MG
APOSOR2270-1GEA
2070
CZK
APOSOR2270-1G
APOSOR2270-250MG
1-Phenylcyclobutanecarboxylic acid
1-Phenylcyclobutanecarboxylic acid
Vzorec:
C₁₀H₁₀O₂ MW: 176.22 g/mol Bod tání: 100…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00019262 CAS číslo: 37828-19-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...