Tisk…

6-Chloro-2-pyridineboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Chloropyridine-2-boronic Acid

OR2262-1G OR2262-250MG
APOSOR2262-1GEA 7180 CZK
APOSOR2262-1G APOSOR2262-250MG
6-Chloro-2-pyridineboronic acid
6-Chloro-2-pyridineboronic acid
Vzorec: C₅H₅BClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD08063028
CAS číslo: 652148-90-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR