Sodium-2-chloro-5-nitrobenzenesulphonate
Dodavatel: Apollo Scientific
OR22597-250MG
OR22597-500MG
OR22597-1G
APOSOR22597-250MGEA
1870
CZK
APOSOR22597-250MG
APOSOR22597-500MG
APOSOR22597-1G
Sodium-2-chloro-5-nitrobenzenesulphonate
Sodium-2-chloro-5-nitrobenzenesulphonate
Vzorec:
C₆H₃ClNNaO₅S MW: 259,6 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00024895 CAS číslo: 946-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...