Tisk…

1-Adamantaneacetic acid 98%

Dodavatel: Apollo Scientific

Synonyms: 2-(Adamantan-1-yl)acetic acid, 1-Carboxymethyladamantane

OR22506-10G OR22506-25G OR22506-5G
APOSOR22506-10GEA 744 CZK
APOSOR22506-10G APOSOR22506-25G APOSOR22506-5G
1-Adamantaneacetic acid 98%
1-Adamantaneacetic acid
Vzorec: C₁₂H₁₈O₂
MW: 194,27 g/mol
Bod tání: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD00074728
CAS číslo: 4942-47-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR