1-Bromoadamantane 98%
Dodavatel: Apollo Scientific
OR22505-100G
OR22505-25G
APOSOR22505-100GEA
1450
CZK
APOSOR22505-100G
APOSOR22505-25G
1-Bromoadamantane 98%
1-Bromoadamantane
Vzorec:
C₁₀H₁₅Br Bod tání: 116…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074721 CAS číslo: 768-90-1 EINECS: 212-200-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...