Norbornane-2-carboxylic acid 99%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Bicyclo[2.2.1]heptane-2-carboxylic acid
OR22442-1G
OR22442-5G
APOSOR22442-1GEA
622
CZK
APOSOR22442-1G
APOSOR22442-5G
Norbornane-2-carboxylic acid 99%
Norbornane-2-carboxylic acid
Vzorec:
C₈H₁₂O₂ MW: 140,18 g/mol Bod varu: 134…136 °C/16mmHg Bod tání: 40 °C Bod vzplanutí: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00167749 CAS číslo: 824-62-4 EINECS: 212-532-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...