4,5-Dibromo-2-thenoic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4,5-Dibromo-2-thiophenecarboxylic acid, 4,5-Dibromothiophene-2-carboxylic acid
OR22419-25G
OR22419-5G
APOSOR22419-25GEA
8070
CZK
APOSOR22419-25G
APOSOR22419-5G
4,5-Dibromo-2-thenoic acid
4,5-Dibromo-2-thenoic acid
Vzorec:
C₅H₂Br₂O₂S MW: 285.94 Bod tání: 225…232 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00039682 CAS číslo: 6324-10-3 EINECS: 228-683-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...