Tisk…

Fmoc-Lys(ME,Boc)-OH

Dodavatel: Apollo Scientific
Varování

Synonyms: Fmoc-Lys(Boc)(Me)-OH, Fmoc-Lys(Me,Boc)-OH

OR2236-100MG OR2236-1G OR2236-250MG
APOSOR2236-100MGEA 662 CZK
APOSOR2236-100MG APOSOR2236-1G APOSOR2236-250MG
Fmoc-Lys(ME,Boc)-OH
Fmoc-Lys(ME,Boc)-OH
Vzorec: C₂₇H₃₄N₂O₆
MW: 482,577 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01861332
CAS číslo: 951695-85-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR