Tisk…

2-Methyl-6-quinolinol 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Hydroxy-2-methylquinoline

OR22357-1G OR22357-5G OR22357-10G
APOSOR22357-1GEA 2280 CZK
APOSOR22357-1G APOSOR22357-5G APOSOR22357-10G
2-Methyl-6-quinolinol 98%
2-Methyl-6-quinolinol
Vzorec: C₁₀H₉NO
MW: 159,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047600
CAS číslo: 613-21-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR