Tisk…

n-Octyl acrylate 95%

Dodavatel: Apollo Scientific

Synonyms: Acrylic acid n-Octyl ester

OR22271-100G OR22271-25G
APOSOR22271-100GEA 10750 CZK
APOSOR22271-100G APOSOR22271-25G
n-Octyl acrylate 95%
n-Octyl acrylate
Vzorec: C₁₁H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00048935
CAS číslo: 2499-59-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR