Tisk…

2-Amino-6-ethylpyridine 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 6-Ethylpyridin-2-amine, 6-Ethyl-2-pyridylamine

OR22212-1G
APOSOR22212-1GEA 622 CZK
APOSOR22212-1G
2-Amino-6-ethylpyridine 95%
2-Amino-6-ethylpyridine
Vzorec: C₇H₁₀N₂
MW: 122,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00218265
CAS číslo: 21717-29-3
EINECS: 244-540-7
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR