Ethyl-5-nitroindole-2-carboxylate 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Ethyl 5-nitro-1H-indole-2-carboxylate
OR2221-10G
OR2221-5G
APOSOR2221-10GEA
2070
CZK
APOSOR2221-10G
APOSOR2221-5G
Ethyl-5-nitroindole-2-carboxylate 98%
Ethyl-5-nitroindole-2-carboxylate
Vzorec:
C₁₁H₁₀N₂O₄ MW: 234,211 g/mol Bod tání: 223…225 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00216477 CAS číslo: 16732-57-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...