Tisk…

Pentaerythrityl tetrabromide 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1,3-Dibromo-2,2-bis(bromomethyl)propane, Tetrakis(bromomethyl)methane, Pentaerythrityltetrabromide

OR22172-100G OR22172-25G
APOSOR22172-100GEA 10880 CZK
APOSOR22172-100G APOSOR22172-25G
Pentaerythrityl tetrabromide 98%
Pentaerythrityl tetrabromide
Vzorec: C₅H₈Br₄
MW: 387.73
Bod varu: 305
Bod tání: 163
Storage Temperature: Ambient
MDL Number: MFCD00000210
CAS číslo: 3229-00-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR