1-(3-Chloro-2-hydroxyphenyl)ethanone
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3'-Chloro-2'-hydroxyacetophenone
OR22015-1G
OR22015-5G
APOSOR22015-1GEA
1540
CZK
APOSOR22015-1G
APOSOR22015-5G
1-(3-Chloro-2-hydroxyphenyl)ethanone
1-(3-Chloro-2-hydroxyphenyl)ethanone
Vzorec:
C₈H₇ClO₂ Bod tání: 47…49 °C Storage Temperature: Freezer |
MDL Number:
MFCD00100492 CAS číslo: 3226-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...