2-Chloro-3,4-dimethoxybenzyl alcohol
Dodavatel: Apollo Scientific
Synonyms:
(2-Chloro-3,4-dimethoxyphenyl)methanol
OR21959-10G
OR21959-1G
OR21959-5G
APOSOR21959-10GEA
7410
CZK
APOSOR21959-10G
APOSOR21959-1G
APOSOR21959-5G
2-Chloro-3,4-dimethoxybenzyl alcohol
2-Chloro-3,4-dimethoxybenzyl alcohol
Vzorec:
C₉H₁₁ClO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00210141 CAS číslo: 93983-13-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...