3,5-Di-tert-butyl-4-hydroxyphenylacetic acid 97%
Dodavatel: Apollo Scientific
OR21938-10G
OR21938-5G
OR21938-25G
APOSOR21938-10GEA
3970
CZK
APOSOR21938-10G
APOSOR21938-5G
APOSOR21938-25G
3,5-Di-tert-butyl-4-hydroxyphenylacetic acid 97%
3,5-Di-tert-butyl-4-hydroxyphenylacetic acid
Vzorec:
C₁₆H₂₄O₃ MW: 264,37 g/mol Bod tání: 157…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00156138 CAS číslo: 1611-03-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...