Tisk…

4-Ethylbenzoic acid

Dodavatel: Apollo Scientific

OR21929-25G OR21929-100G OR21929-250G
APOSOR21929-25GEA 622 CZK
APOSOR21929-25G APOSOR21929-100G APOSOR21929-250G
4-Ethylbenzoic acid
4-Ethylbenzoic acid
Vzorec: C₉H₁₀O₂
MW: 150,18 g/mol
MDL Number: MFCD00002570
CAS číslo: 619-64-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR