Tisk…

1-Bromo-4-heptylbenzene

Dodavatel: Apollo Scientific

Synonyms: (4-Bromophenyl)heptane

OR21924-5G
APOSOR21924-5GEA 2200 CZK
APOSOR21924-5G
1-Bromo-4-heptylbenzene
1-Bromo-4-heptylbenzene
Vzorec: C₁₃H₁₉Br
MW: 255,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061115
CAS číslo: 76287-49-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR