4-Pentylbenzoic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Amylbenzoic acid, 4-n-Pentylbenzoic acid, 4-Pentylbenzoic acid
OR21922-25G
OR21922-5G
APOSOR21922-25GEA
1040
CZK
APOSOR21922-25G
APOSOR21922-5G
4-Pentylbenzoic acid 97%
4-Pentylbenzoic acid
Vzorec:
C₁₂H₁₆O₂ MW: 192,26 g/mol Bod varu: 261 °C Bod tání: 87…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002572 CAS číslo: 26311-45-5 EINECS: 247-607-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...