Tisk…

4-(4-Octylphenyl)-4-oxobutanoic acid

Dodavatel: Apollo Scientific

OR21909-10G OR21909-1G OR21909-5G
APOSOR21909-10GEA 24600 CZK
APOSOR21909-10G APOSOR21909-1G APOSOR21909-5G
4-(4-Octylphenyl)-4-oxobutanoic acid
4-(4-Octylphenyl)-4-oxobutanoic acid
Vzorec: C₁₈H₂₆O₃
Storage Temperature: Ambient
MDL Number: MFCD00173760
CAS číslo: 64779-10-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR