Pentachlorobenzonitrile
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,3,4,5,6-Pentachlorobenzonitrile
OR21863-1G
OR21863-5G
APOSOR21863-1GEA
622
CZK
APOSOR21863-1G
APOSOR21863-5G
Pentachlorobenzonitrile
Pentachlorobenzonitrile
Vzorec:
C₇Cl₅N MW: 275.35 Storage Temperature: Refrigerator |
MDL Number:
MFCD00173666 CAS číslo: 20925-85-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...