Tisk…

Chlorodiphenylphosphine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: DPPC, Diphenylchlorophosphine

OR2181-100G OR2181-25G
APOSOR2181-100GEA 2070 CZK
APOSOR2181-100G APOSOR2181-25G
Chlorodiphenylphosphine
Chlorodiphenylphosphine
Vzorec: C₁₂H₁₀ClP
Bod varu: 320 °C
Hustota: 1,229 g/cm³ (25 °C)
Bod vzplanutí: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000529
CAS číslo: 1079-66-9
EINECS: 214-093-2
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR