Tisk…

3-(2-Chloro-3,4-dimethoxyphenyl)acryloyl chloride

Dodavatel: Apollo Scientific

Synonyms: 3-(2-chloro-3,4-dimethoxyphenyl)prop-2-enoyl chloride

OR21791-1G OR21791-250MG OR21791-500MG OR21791-5G
APOSOR21791-1GEA 7440 CZK
APOSOR21791-1G APOSOR21791-250MG APOSOR21791-500MG APOSOR21791-5G
3-(2-Chloro-3,4-dimethoxyphenyl)acryloyl chloride
3-(2-Chloro-3,4-dimethoxyphenyl)acryloyl chloride
Vzorec: C₁₁H₁₀Cl₂O₃
MW: 261,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00084949
CAS číslo: 175136-00-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR