Tisk…

2,7-Dimethoxynaphthalene 98%

Dodavatel: Apollo Scientific

OR2179-100G OR2179-25G
APOSOR2179-100GEA 1920 CZK
APOSOR2179-100G APOSOR2179-25G
2,7-Dimethoxynaphthalene 98%
2,7-Dimethoxynaphthalene
Vzorec: C₁₂H₁₂O₂
Bod tání: 137…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00004064
CAS číslo: 3469-26-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR