Tisk…

(3-Methoxyphenyl)acetic acid ethyl ester

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl (3-methoxyphenyl)acetate, Ethyl 2-(3-methoxyphenyl)acetate

OR21783-25G OR21783-5G OR21783-100G
APOSOR21783-25GEA 2110 CZK
APOSOR21783-25G APOSOR21783-5G APOSOR21783-100G
(3-Methoxyphenyl)acetic acid ethyl ester
(3-Methoxyphenyl)acetic acid ethyl ester
Vzorec: C₁₁H₁₄O₃
MW: 194.23
Bod varu: 109…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00026897
CAS číslo: 35553-92-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR