4-[1-(4-Aminophenyl)-1-methylethyl]aniline
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4,4'-(Propane-2,2-diyl)dianiline
OR21754-5G
APOSOR21754-5GEA
7910
CZK
APOSOR21754-5G
4-[1-(4-Aminophenyl)-1-methylethyl]aniline
4-[1-(4-Aminophenyl)-1-methylethyl]aniline
Vzorec:
C₁₅H₁₈N₂ MW: 226,32 g/mol Bod tání: 126…133 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00438683 CAS číslo: 2479-47-2 EINECS: 219-612-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...