Tisk…

4-Chloro-5-methoxy-2-methylpyridazin-3(2H)-one

Dodavatel: Apollo Scientific
Varování

OR21746-1G OR21746-250MG OR21746-500MG
APOSOR21746-1GEA 10820 CZK
APOSOR21746-1G APOSOR21746-250MG APOSOR21746-500MG
4-Chloro-5-methoxy-2-methylpyridazin-3(2H)-one
4-Chloro-5-methoxy-2-methylpyridazin-3(2H)-one
Vzorec: C₆H₇ClN₂O₂
MW: 174,59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00067854
CAS číslo: 14628-57-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR