3-Bromo-4-chlorophenol 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR2161-5G
OR2161-100G
OR2161-25G
APOSOR2161-5GEA
870
CZK
APOSOR2161-5G
APOSOR2161-100G
APOSOR2161-25G
3-Bromo-4-chlorophenol 98%
3-Bromo-4-chlorophenol
Vzorec:
C₆H₄BrClO MW: 207.4500 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00070740 CAS číslo: 13659-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...