Tisk…

(2-Bromo-5-chlorophenyl)acetic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(2-Bromo-5-chlorophenyl)acetic acid

OR2153-100G OR2153-10G OR2153-25G
APOSOR2153-100GEA 15510 CZK
APOSOR2153-100G APOSOR2153-10G APOSOR2153-25G
(2-Bromo-5-chlorophenyl)acetic acid
(2-Bromo-5-chlorophenyl)acetic acid
Vzorec: C₈H₆BrClO₂
MW: 249,4900 g/mol
Bod tání: 101…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00153890
CAS číslo: 81682-38-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR