Tisk…

2-Methyl-2H-indazol-4-amine 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Amino-2-methyl-2H-indazole

OR2152-1G OR2152-250MG
APOSOR2152-1GEA 3580 CZK
APOSOR2152-1G APOSOR2152-250MG
2-Methyl-2H-indazol-4-amine 95%
2-Methyl-2H-indazol-4-amine
Vzorec: C₈H₉N₃
MW: 147,1800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09870044
CAS číslo: 82013-51-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR