Tisk…

6-Hydroxypyridazine-3-carboxylic acid monohydrate

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Hydroxy-3-pyridazinecarboxylic Acid Monohydrate, 1,6-Dihydro-6-oxopyridazine-3-carboxylic acid monohydrate

OR21502-10G OR21502-1G OR21502-5G
APOSOR21502-10GEA 1040 CZK
APOSOR21502-10G APOSOR21502-1G APOSOR21502-5G
6-Hydroxypyridazine-3-carboxylic acid monohydrate
6-Hydroxypyridazine-3-carboxylic acid monohydrate
Vzorec: C₅H₆N₂O₄
Bod tání: 246…247 °C
Storage Temperature: Ambient
MDL Number: MFCD09064936
CAS číslo: 306934-80-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR