Methylcyclopentadiene dimer 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR2150-1KG
OR2150-250G
OR2150-100G
APOSOR2150-1KGEA
4370
CZK
APOSOR2150-1KG
APOSOR2150-250G
APOSOR2150-100G
Methylcyclopentadiene dimer 95%
Methylcyclopentadiene dimer
Vzorec:
C₁₂H₁₆ MW: 160,26 g/mol Bod varu: 198…201 °C Bod tání: –51 °C Hustota: 0,941 g/cm³ (25 °C) Bod vzplanutí: 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082297 CAS číslo: 26472-00-4 EINECS: 247-724-5 UN: 3295 ADR: 3,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...