Tisk…

2-(Phenylsulphanyl)nicotinonitrile

Dodavatel: Apollo Scientific

Synonyms: 2-(Phenylthio)nicotinonitrile

OR21429-10G OR21429-1G OR21429-5G
APOSOR21429-10GEA 12340 CZK
APOSOR21429-10G APOSOR21429-1G APOSOR21429-5G
2-(Phenylsulphanyl)nicotinonitrile
2-(Phenylsulphanyl)nicotinonitrile
Vzorec: C₁₂H₈N₂S
MW: 212,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052659
CAS číslo: 35620-68-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR