3,4,5-Tribromo-1H-pyrazole 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3,4,5-Tribromopyrazole
OR21421-100G
OR21421-25G
OR21421-5G
APOSOR21421-100GEA
3270
CZK
APOSOR21421-100G
APOSOR21421-25G
APOSOR21421-5G
3,4,5-Tribromo-1H-pyrazole 98%
3,4,5-Tribromo-1H-pyrazole
Vzorec:
C₃HBr₃N₂ MW: 304,78 g/mol Bod tání: 184…188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040248 CAS číslo: 17635-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...