Thieno[2,3-c]pyridine
Dodavatel: Apollo Scientific
|
Varování
|
OR2141-1G
OR2141-250MG
OR2141-5G
APOSOR2141-1GEA
1660
CZK
APOSOR2141-1G
APOSOR2141-250MG
APOSOR2141-5G
Thieno[2,3-c]pyridine
Thieno[2,3-c]pyridine
Vzorec:
C₇H₅NS Storage Temperature: Refrigerator |
MDL Number:
MFCD01760492 CAS číslo: 272-12-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...