Tisk…

Ethyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylate

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 7-azaindole-2-carboxylate, Ethyl 1H-pyrrolo[2,3-b]pyridine-2-carboxylate

OR2140-250MG OR2140-100MG
APOSOR2140-250MGEA 953 CZK
APOSOR2140-250MG APOSOR2140-100MG
Ethyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylate
Ethyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylate
Vzorec: C₁₀H₁₀N₂O₂
MW: 190,2000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09998715
CAS číslo: 221675-35-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR