2-(1H)-Tetrahydropyrimidinethione
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Tetrahydropyrimidine-2(1H)-thione
OR21392-100G
OR21392-5G
OR21392-25G
APOSOR21392-100GEA
7440
CZK
APOSOR21392-100G
APOSOR21392-5G
APOSOR21392-25G
2-(1H)-Tetrahydropyrimidinethione
2-(1H)-Tetrahydropyrimidinethione
Vzorec:
C₄H₈N₂S MW: 116.19 g/mol Bod tání: 210…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060526 CAS číslo: 2055-46-1 EINECS: 218-152-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...