Ethyl-4-amino-2-mercaptopyrimidine-5-carboxylate
Dodavatel: Apollo Scientific
OR21285-5G
APOSOR21285-5GEA
1740
CZK
APOSOR21285-5G
Ethyl-4-amino-2-mercaptopyrimidine-5-carboxylate
Ethyl-4-amino-2-mercaptopyrimidine-5-carboxylate
Vzorec:
C₇H₉N₃O₂S MW: 199,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00023227 CAS číslo: 774-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...