3,5-Dibromosalicylaldehyde
Dodavatel: Apollo Scientific
Synonyms:
3,5-Dibromo-2-hydroxybenzaldehyde
OR21193-100G
OR21193-500G
APOSOR21193-100GEA
1660
CZK
APOSOR21193-100G
APOSOR21193-500G
3,5-Dibromosalicylaldehyde
3,5-Dibromosalicylaldehyde
Vzorec:
C₇H₄Br₂O₂ MW: 279,92 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003318 CAS číslo: 90-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...