1-(4-Nitrophenyl)imidazolidin-2-one
Dodavatel: Apollo Scientific
OR2115-1G
OR2115-250MG
APOSOR2115-1GEA
9720
CZK
APOSOR2115-1G
APOSOR2115-250MG
1-(4-Nitrophenyl)imidazolidin-2-one
1-(4-Nitrophenyl)imidazolidin-2-one
Vzorec:
C₉H₉N₃O₃ Bod tání: 245 °C Storage Temperature: Ambient |
MDL Number:
MFCD09998748 CAS číslo: 89518-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...