2'-Amino-4'-methoxyacetanilide
Dodavatel: Apollo Scientific
|
Varování
|
OR21097-1G
OR21097-5G
APOSOR21097-1GEA
0
CZK
APOSOR21097-1G
APOSOR21097-5G
2'-Amino-4'-methoxyacetanilide
2'-Amino-4'-methoxyacetanilide
Vzorec:
C₉H₁₂N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00026124 CAS číslo: 5472-37-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...