2,5-Dichloro-3-nitropyridine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR2086-5G
OR2086-10G
APOSOR2086-5GEA
622
CZK
APOSOR2086-5G
APOSOR2086-10G
2,5-Dichloro-3-nitropyridine 97%
2,5-Dichloro-3-nitropyridine
Vzorec:
C₅H₂Cl₂N₂O₂ Bod tání: 39…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD06658963 CAS číslo: 21427-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...